Multi-Parameter Meter, Handheld, pH/Oxi 340i
Supplier: Avantor
Certificates
About this item
These robust pocket instruments for pH and oxygen measurements are water resistant and fulfill the requirements of IP 67.
All instruments have a large, easy to read display (60×45 mm), simultaneously showing oxygen measurement and temperature. Other sets available on request.
Ordering information: pH/Oxi set includes instrument, professional case, pH and oxygen electrodes, holder STH 320 and accessories.