Order Entry
Puerto Rico
ContactUsLinkComponent
Accessories for Graduated Fleakers, Miscellaneous Laboratory Glassware, Quality Lab Accessories
Accessories for Graduated Fleakers, Miscellaneous Laboratory Glassware, Quality Lab Accessories
Catalog # 89133-020
Supplier:  QLA
undefined
Accessories for Graduated Fleakers, Miscellaneous Laboratory Glassware, Quality Lab Accessories
Catalog # 89133-020
Supplier:  QLA
Supplier Number:  FLEAKCAP-UN

Specifications

  • Description:
    Fleaker Cap fits 150 mL, 300 mL and 500 mL Fleakers
  • Cat. No.:
    89133-020
  • Supplier no.:
    FLEAKCAP-UN

Specifications

About this item