ContactUsLinkComponent
Cotton Tipped Applicator Swab, Dynalon
Cotton Tipped Applicator Swab, Dynalon
Catalog # 30621-018
Supplier:  Dynalon
undefined
Cotton Tipped Applicator Swab, Dynalon
Catalog # 30621-018
Supplier:  Dynalon
Supplier Number:  394365
Specifications
  • Length:
    6"
  • Material:
    Wood
  • Cat. No.:
    30621-018
  • Supplier no.:
    394365
Specifications
About this item
Disposable cotton swabs manufactured of USP grade purified cotton firmly attached to one end of wood shaft.
May be autoclaved. Useful in bacteriology labs for inoculating broths, plated media or for obtaining culture specimens. Also useful for cleaning delicate instruments.