VWR® Pipette Tips, Extra Long
Catalog # 89368-966
Supplier: VWR International
Specifications
- Volume:10 µl
- Environmentally Preferable:Sustainable Packaging,Low Manufacturing Impact
- Tip color:Clear
- Packed:Bulk, 1000/Bag, 10 Bag/Case
- Cat. No.:89368-966
- Tip style:Extra Long Pipette Tips
Specifications
About this item
These polypropylene tips are longer than most 10 µl tips to allow users to reach down into 0.65 ml tubes and multiwell plates.
- RNase/DNase-, endotoxin-, protease-free, ATP-free, BPA -fee
- Tips and racks are autoclavable
- Supplied in bags or refillable racks
Tips feature a fine thin capillary section at the end that is non bevelled and very flexible; enables users to dispense the last small amount of sample from the tip.