Anti-IgG1 gamma Rabbit Polyclonal Antibody (Cy3®)
RL610-404-040
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G1 gamma
- Antigen Symbol:IgG1 gamma
- Clonality:Polyclonal
- Conjugation:Cy3®
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Size:500 µL
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:Mouse IgG1 heavy chain
- Tested Applications:Fluoresence ELISA, Fluoresence Western Blot
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL610-404-040
- Supplier no.:610-404-040
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG1 gamma
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse