ContactUsLinkComponent
ErgoPerfect® Ergonomic Sit/Stand Stool, ERGOMAT®
  10054-358
 :  Ergomat
undefined
ErgoPerfect® Ergonomic Sit/Stand Stool, ERGOMAT®
  10054-358
 :  Ergomat
 :  ERGO600S
  • Description:
    Ergonomic Sit/Stand Stool
  • Model:
    10054-358
  • Cat. No.:
    10054-358
  • Seat height:
    22 to 31.5"
  • Supplier no.:
    ERGO600S
The ErgoPerfect® Sit/Stand is a robustly constructed, lightweight ergonomic stool that is perfect for use in a range of working environments, from lab benches to customer service counters
The saddle tilts between 3° and 11° at a height which ranges between 22" and 31.5". Additional height can be achieved with optional taller glides if necessary. The soft, durable polyurethane upholstery has a bubble texture to promote circulation, and to provide grip when the saddle is pitched forward. At 19 pounds, the stool is easily moved between work stations by using the handle that is molded as part of the back of the saddle.