Order Entry
Puerto Rico
ContactUsLinkComponent
 
1,1-Dimethylbiguanide hydrochloride, Sigma-Aldrich®
  80016-622
 :  
1,1-Dimethylbiguanide hydrochloride, Sigma-Aldrich®
  80016-622
 :  317240-5GM
 :  

 

  • Size:
    5 g
  • Cat. No.:
    80016-622
  • Supplier no.:
    317240-5GM

 

 

  • Molecular Weight:
    165.6

 

 

 :
 :165.625 g/mol
 :1115-70-4
 :C₄H₁₂ClN₅
 :MFCD00012582