Thin Chromatography Paper, Grade 601, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 601 is a thin chromatography paper with medium flow rate and a smooth uniform white surface.
Specifications
- Catalog No:
- 490005-080
- 47745-904
- 47749-844
- 490005-082
- 47745-906
- 47745-898
- 490005-084
- 28334-194
- Thickness:
- 0.19 mm
- 0.19 mm
- 0.19 mm
- 0.19 mm
- 0.19 mm
- 0.19 mm
- 0.19 mm
- 0.19 mm
- Absorption Speed:
- 125 mm / 30 min
- 125 mm / 30 min
- 125 mm / 30 min
- 125 mm / 30 min
- 125 mm / 30 min
- 125 mm / 30 min
- 125 mm / 30 min
- 125 mm / 30 min