- Description:Model C2-S System
- Cat. No.:27373-322
- Supplier no.:C2-S
Horizontal microgel system is designed for quick restriction fragment analysis
Glass slides measuring 2x3" (5x7.5cm) serve as the gel tray. To use, pour and form thin gels by surface tension directly on glass slide; complete run lasts five to 10 minutes. Using available caster and casting slides, gels can be cast and solidified in less than 10 minutes.
Microgel may also be used to quickly check a few PCR samples before loading a large number of samples on a larger gel, saving time.