ContactUsLinkComponent
SP Bel-Art Wash Bottle Carrier, Bel-Art Products, a part of SP
SP Bel-Art Wash Bottle Carrier, Bel-Art Products, a part of SP
Catalog # 490012-274
undefined
SP Bel-Art Wash Bottle Carrier, Bel-Art Products, a part of SP
Catalog # 490012-274
Supplier Number:  F11640-0001
Specifications
  • Description:
    Wash Bottle Carrier
  • Cat. No.:
    490012-274
  • Supplier no.:
    F11640-0001
Specifications
About this item
This sturdy, lightweight polypropylene carrier helps personnel safely and conveniently transport up to six wash bottles in one convenient holder.
  • Sturdy design
  • Lightweight
  • Polypropylene construction
  • Conveniently transports up to six wash bottles
  • Chemically resistant
Its secure handle affords both comfort and control. Autoclavable at 121 °C (250 °F) and chemical resistant. This carrier holds either 8 oz. or 16 oz. bottles. Dimensions are 286 x 194 x 225 mm high (11-1/4 x 7-5/8 x 8-7/8"). Bottles are not included with the carrier.