Order Entry
Puerto Rico
ContactUsLinkComponent
 
SP Bel-Art Double Eyewash Station, Bel-Art Products, a part of SP
  89522-668
undefined
SP Bel-Art Double Eyewash Station, Bel-Art Products, a part of SP
  89522-668
 :  F24868-0000

 

  • Description:
    Double eye wash station, with two empty 32 oz. bottles
  • Cat. No.:
    89522-668
  • Supplier no.:
    248680000

 

 

Supplied with two empty 32 oz.(1000 ml) bottles, this station provides a half-gallon of solution capacity.

  • Bottle dust covers keep out contamination
  • Eyewash solution not included

The vacuum-formed backboard has two bottle wells, a mirror, 4 screw holes for mounting, and full instructions for use and maintenance. 18×14" (46×36 cm).

 : For full sterile saline bottles, see 30617-158 series.