ContactUsLinkComponent
MBP 1 to 300 µl Graduated Pipette Tips, Thermo Scientific
MBP 1 to 300 µl Graduated Pipette Tips, Thermo Scientific
Catalog # 490015-830
Supplier:  Thermo Scientific
MBP 1 to 300 µl Graduated Pipette Tips, Thermo Scientific
Catalog # 490015-830
Supplier:  Thermo Scientific
Supplier Number:  108-96RG
Specifications
  • Volume:
    1 - 300 µl
  • Packed:
    96 Tip Rack
  • Cat. No.:
    490015-830
  • Tip style:
    Graduated
  • Supplier no.:
    108-96RG
Specifications
About this item
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Titertek®, Biohit Proline®, Costar®, Finnpipette®, Jencons Sealpette®, Labpette™, Nichiryo, Oxford-Benchmate®, Pipetman®, and GeneMate®. Natural color.
A mold release agent is NOT used in the manufacturing of these pipette tips. Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request. Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.