ContactUsLinkComponent
Diversified Biotech Synergel™ Agarose Additive
Diversified Biotech Synergel™ Agarose Additive
Catalog # 52857-970
Small Business Enterprise
undefined
Diversified Biotech Synergel™ Agarose Additive
Catalog # 52857-970
Supplier Number:  SYN-100
Small Business Enterprise
Specifications
  • Description:
    Synergel Agarose Additive
  • Cat. No.:
    52857-970
  • Supplier no.:
    SYN100
Specifications
About this item
Better sieving than agarose, 1 gram Synergel = 2 grams agarose.High Resolution
Synergel/Agarose gels exhibit improved spatial separation and tighter banding for DNA fragments of 50-2000bp. No Background Fluorescence. The superior optical clarity permits high quality photo-documentation. Increased Load Capacity—Accommodates greater quantities of DNA reducing band tailing and smearing. Easy To Use; mix, boil, and pour.