ContactUsLinkComponent
Accessories for Labnet Z206A Digital High Capacity Compact Universal Centrifuge, Labnet International
Accessories for Labnet Z206A Digital High Capacity Compact Universal Centrifuge, Labnet International
Catalog # 490008-686
undefined
Accessories for Labnet Z206A Digital High Capacity Compact Universal Centrifuge, Labnet International
Catalog # 490008-686
Supplier Number:  C0200-1
Specifications
  • Description:
    Angle Rotor, 18 x 1.5/2.0 mL (max. 6,000 rpm/2,390 x g)
  • Cat. No.:
    490008-686
  • Supplier no.:
    C0200-1