Order Entry
Puerto Rico
ContactUsLinkComponent
pH Singles Calibration Pouches, Oakton Instruments
pH Singles Calibration Pouches, Oakton Instruments
Catalog # 101076-328
CAS Number:  
undefined
pH Singles Calibration Pouches, Oakton Instruments
Catalog # 101076-328
Supplier Number:  WD-35653-04
CAS Number:  

Specifications

  • Description:
    pH Singles Calibration Pouches
  • Cat. No.:
    101076-328
  • Supplier no.:
    WD-35653-04

Specifications

About this item

Calibrate your pH meter in the lab or in the field with convenient, portable, and highly accurate pH Singles

  • No Unwieldy Bottles to Carry or Clean

The pouches are airtight for maximum freshness and unquestionable reliability. All solutions are certified traceable to NIST Standard Reference Materials for the highest accuracy. You get twenty 20 mL pouches, five each of pH 4, 7, and 10, and rinse water.