SP Bel-Art Cryo-Safe™ Cold Box, Bel-Art Products, a part of SP
490012-162
- Description:Cryo-Safe Cold Box
- Cat. No.:490012-162
- Supplier no.:F18847-0002
Use this compact cooler on the benchtop to maintain twelve 1.5 mL centrifuge tubes at -15°C for 1.5 hours, or below 0°C for up to 4 hours eliminating ice baths or trips to the freezer.
- Prevents expensive enzymes and critical samples from warming
- Active by placing it in a -20°C freezer for 24 hours
- Stackable
Units have a lid to protect the contents and can be stacked to save space during storage or transport.