Biochrom Cuvettes for Photometers and Spectrophotometers
Supplier: BIOCHROM US BE
Certificates
About this item
Quartz glass cuvettes are suitable for use in the wavelength range 230 - 2500 nm. Optical glass cuvettes are suitable for use in the wavelength range 320 - 2500 nm.
- Sizes range from 1.4 ml to 750 µl volume
Biochrom has a wide range of cuvettes for numerous spectrophotometers and photometers. Ranging from quartz, plastic, optical glass or acrylic, and various volume sizes, you are sure to find what you need from this selection of cuvettes!
Ordering information: Quartz (490005-884, 490005-912), plastic (490014-202, 490005-866), optical glass (490014-208), acrylic (490014-204). Pack of 100 (490005-886, 490014-202, 490014-204). 10 mm (490014-202). semi-micro (490014-202, 490005-886) or ultra micro (490005-912).