Order Entry
Puerto Rico
ContactUsLinkComponent
 
GeneMate Mini Dry Bath with Heated Lid
Catalog # 490000-072
Supplier:  Ward's
undefined
GeneMate Mini Dry Bath with Heated Lid
Catalog # 490000-072
Supplier:  Ward's
Supplier Number:  BSH200-HL-GM

Specifications

  • Temperature Range:
    Ambient +5 to 100 °C
  • Description:
    Mini Dry Bath with Heated Lid
  • Temperature Accuracy:
    +/- 0.3 °C
  • Electrical:
    Universal Voltage, 100 to 240V, 50-60Hz, 50W
  • W×D×H:
    10.5×15×11.5 cm (4×6×4.5")
  • Chamber Dimensions:
    4.7 x 7.1 x 3.2 cm (1.85 x 2.8 x 1.25")
  • Uniformity:
    +/- 0.2 °C
  • Weight:
    1 kg (2 lbs.)
  • Increments:
    0.1°C
  • Cat. No.:
    490000-072
  • Supplier no.:
    BSH200-HL-GM

Specifications

About this item

This Mini Dry Bath with Heated Lid is the first mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation.

  • 2-year full replacement warranty
  • Heated lid prevents evaporation & condensation
  • Compact, fits in the palm of your hand
  • Independent temperature control of lid & block
  • Exchangeable blocks for 0.2 to 0.5 mL tubes
  • Room temperature to 37°C in just 2 minutes
  • Blocks Sold Separately

Both the block and lid temperature can be selected independently in increments of 0.1°C, making this the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions & other temperature hyper-sensitive procedures.