- Description:Premium Multi-Gen 7XL Homogenizing Package (includes Bio-Gen PRO200 and 12-pack of Multi-Gen 7XL Generator Probes)
- Cat. No.:490009-688
- Supplier no.:PK-01200PMGXL
The Multi-Gen is ideal for multiple sample processing, where cross-contamination could be a concern. The Multi-Generator pack allows users to homogenize 12 or 24 samples (depending upon which package is purchased) at a time, before users have to worry about cleaning the generator and the adapter. This allows for hands-free ejection of the used generator after each use. Multi-Gen generators are approximately 7x75mm, with the ideal tube size for use with the Multi-Gen system being a 1.5/2.0mL microtube or a 5mL tube.
PRO Scientific's Multi-Gen 7XL is a newly redesigned version of the Multi-Gen 7. With this new design, users can now effectively process tougher, bigger samples without worrying about cross-contamination. The new Multi-Gen 7XL features an open slotted tooth design, making these generators ideal for tougher samples. Additionally, a longer shaft allows easy processing in 1.5/2mL microtubes up to 50mL conical tubes. All Multi-Gen generators quickly lock into the special Multi-Gen Motor Unit Adapter. When homogenization is complete, hands-free ejection of the generator minimizes the chance of accidental contamination. The Multi-Pack design reduces cross contamination risk by allowing users to process each sample with a clean generator, and eliminate cleaning between samples. It's 316SS design allows for the utmost in chemical compatibility and all generators can be easily autoclaved at the end of processing.
All packages include everything needed to begin homogenizing in a convenient carrying case.
PRO Scientific's Multi-Gen 7XL is a newly redesigned version of the Multi-Gen 7
With this new design, users can now effectively process tougher, bigger samples without worrying about cross-contamination. The new Multi-Gen 7XL features an open slotted tooth design, making these generators ideal for tougher samples. Additionally, a longer shaft allows easy processing in 1.5/2mL microtubes up to 50mL conical tubes. All Multi-Gen generators quickly lock into the special Multi-Gen Motor Unit Adapter. When homogenization is complete, hands-free ejection of the generator minimizes the chance of accidental contamination. The Multi-Pack design reduces cross contamination risk by allowing users to process each sample with a clean generator, and eliminate cleaning between samples. It's 316SS design allows for the utmost in chemical compatibility and all generators can be easily autoclaved at the end of processing.