Sonics 500 and 750-Watt Ultrasonic Processors
Supplier: SONICS & MATERIALS BE
Certificates
About this item
Ultrasonic processors process a wide range of organic and inorganic materials, from 250 ml to 1 L (actual processing volume of a specific probe or accessory is application specific).
- Programmable
- Variable power output
- 10 hour timer
- Pulse mode
- Wattage and energy display
Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing.
The 750 W models have the capability of operating the largest assortment of horns and accessories.
The VCX 750 model also includes temperature monitoring and energy setpoint programming.
Ordering information: Temperature probe, stand (with clamp), and other accessories are not included. Many optional accessories are available to meet the needs of any application. Contact your Avantor representative for more information.
Delivery: Includes a standard ½” ⌀ probe with replaceable tip and tool kit.
Caution: Do not use a probe with replaceable tip when processing samples containing organic solvents or low surface tension liquids. Use a solid probe instead. Unless otherwise requested, the probe supplied will have a replaceable tip. Contact your Avantor representative for more information.
Specifications
- Catalog No:
- 490010-590
- 490010-580
- Frequency:
- 20 kHz
- 20 kHz
- W×D×H:
- 190×340×235 mm (7.5×13.5×9.25”)
- 190×340×235 mm (7.5×13.5×9.25”)