Weigh-Better Biodegradable Boats, Diversified Biotech
Supplier: Diversified Biotech
New Product
Certificates
About this item
Glassine paper is free of static electricity that plagues disposable plastic weighing dishes (polystyrene and polyethylene).
- No Static Build-Up
- Biodegradable- Guilt-free glassine paper will completely breakdown when discarded
- Contaminant Free- Pure glassine is an ultra-clean "GREEN" paper product
- Easy to Use- Robust and durable yet easy to fold for pouring
- Ideally Sized- Measures 3.5" diameter for standard weighing balances
- Color Contrast- Chocolate-colored glassine reveals every particle of material