Microscope Slide Boxes
Heavy-duty, two-piece microscope slide boxes are ideal for storing or transportating standard slides, 75×25 mm (3×1").
- Designed to securely store slides in grooved slots
- Molded feet secure stacking for easy transport and storage
Closes securely with a rust-resistant nickel-plated clasp and pin hinges. ABS plastic boxes available with either cork or foam lining.
Heavy-duty, two-piece microscope slide boxes are ideal for storing or transportating standard slides, 75×25 mm (3×1").
- Designed to securely store slides in grooved slots
- Molded feet secure stacking for easy transport and storage
Closes securely with a rust-resistant nickel-plated clasp and pin hinges. ABS plastic boxes available with either cork or foam lining.
Product Family Options
- DescriptionCapacityDimensionsColor
- Microscope slide box, lining: cork, lock type: nickel-plated clasp100 Slides208×175×34 mmWhiteEach$24.10Case of 72$1,560.71: HS15994E
- Microscope slide box, lining: cork, lock type: nickel-plated clasp100 Slides208×175×34 mmRedEach$22.06Case of 72$1,578.88: HS15994C
- Microscope slide box, lining: cork, lock type: nickel-plated clasp100 Slides208×175×34 mmBlueEach$28.47Case of 72$1,627.53: HS15994A
- Microscope slide box, lining: cork, lock type: nickel-plated clasp100 Slides208×175×34 mmGreenEach$24.40Case of 72$1,437.33: HS15994B
- Microscope slide box, lining: cork, lock type: nickel-plated clasp100 Slides208×175×34 mmYellowEach$29.41Case of 72$2,013.58: HS15994D
- Microscope slide box, lining: cork, lock type: thumb-latch50 Slides209×86×35 mmRedEach$23.32Case of 72$914.60: HS15996B
- Microscope slide box, lining: cork, lock type: thumb-latch50 Slides209×86×35 mmWhiteEach$31.06Case of 72$1,360.54: HS15996C
- Microscope slide box, lining: foam, lock type: friction fit25 Slides141×88×35 mmWhiteEach$34.42Case of 72$2,475.60: HS15989R
- Microscope slide box, lining: cork, lock type: thumb-latch50 Slides209×86×35 mmBlueEach$41.65Case of 72$1,794.90: HS15996A