Diversified Biotech Wash-N-Dry™ Slide Rack
Catalog # 490007-154
Supplier: Diversified Biotech
Specifications
- Description:Wash-N-Dry Slide Rack
- Cat. No.:490007-154
- Supplier no.:WSDR-2000
Specifications
About this item
Securely holds twelve slides for convenient cleaning, treating, staining and drying. Fits standard slide sizes including 25 mm & 1".
- Offset design allows liquid and air to flow around and between all the slides ensuring best cleaning and faster drying times
- Chemically resistant
- Autoclavable