ContactUsLinkComponent
SP Bel-Art Glass Disposal Box, Bel-Art Products, a part of SP
SP Bel-Art Glass Disposal Box, Bel-Art Products, a part of SP
  490005-580
undefined
SP Bel-Art Glass Disposal Box, Bel-Art Products, a part of SP
  490005-580
 :  F24653-0001
  • Description:
    Glass Disposal Box for Floor
  • Dimensions:
    30 x 30 x 69 cm (12 x 12 x 27")
  • Environmentally Preferable:
  • Cat. No.:
    490005-580
  • Supplier no.:
    F24653-0001
This Glass Disposal Box is a disposable, highly visible receptacle that helps reduce injuries by maintaining a safe laboratory work area.
  • Disposable
  • Highly visible with eye-catching graphics
  • Thick polyethylene construction
The receptacle is supplied with a 2 mil thick polyethylene bag to safely contain broken glass fragments and spilled liquids. When the box is full, the safety cap is pulled into place and the entire unit is ready for disposal. Printed with eye-catching graphics in four languages (English, German, French, Spanish)
 : Packaged flat to save space and shipping costs and is quickly assembled. Available in two sizes: The floor model holds 18 kg (40 lbs) of waste, while the benchtop model holds 5.5kg (12 lbs)