GeneMate Graduated Microcentrifuge Tubes, 1.7 ml, Scientific Specialties
Microcentrifuge tubes features wide cap flange for easy opening, ultimate in clarity and glass-like positive seal design.
- Suitable for temperature range from –90 to 120 °C
- Rated to 20000 ×g
- Ideal for boiling applications
- Free of detectable DNase, RNase, and human DNA
: 500/Pack. Cases come with 10 packs