GeneMate 384-Well PCR Plates, Full-Skirted, One-Notch (A24) Type, Scientific Specialties
Catalog # 490003-838
Supplier: GeneMate
Specifications
- No. of wells:384
- Skirt:Fully skirted
- Well Color:Natural
- Description:PCR plate, one-notch type
- Cat. No.:490003-838
- Supplier no.:3435-00-520IS
Specifications
About this item
Compatible with most commercially available thermal cyclers.
- Ultra-thin wall for optimum heat transfer and higher yield
- High contrast black alphanumerics for rapid sample identification
- Plates can be optimally sealed with real-time sealing film
- Crystal-clear clarity
- Easy to visualize content of each well
- Certified free from DNase, RNase, and detectable DNA
- Also available either pre-barcoded or custom-barcoded for convenient sample identification and tracking