Diversified Biotech Reagent Reservoir Cooler
Supplier: Diversified Biotech
Certificates
About this item
Reagents are kept at ~5°C for approximately 3 hours.
No more messy ice baths, frozen nontoxic gel is contained in a sealed molded cavity. Extends safe working time temperature sensitive reagents. Compatible with 100 and 25 mL reservoirs.