Adjustable Well Plate Stand, Diversified Biotech
490007-136
- Description:Adjustable Well Plate Stand
- Cat. No.:490007-136
- Supplier no.:WPAJ-1000
Achieves accurate pipetting by allowing the flexibility to adjust the angle of the microplate (45° max. tilt).
The stand takes up little benchtop space measuring only 6"L x 4.50"W x 2.50"H.