Cameo 30 mm Syringe Filter, GVS Filter Technology
Catalog # 490007-506
Supplier: GVS FILTER TECHNOLOGY
Small Business Enterprise
Specifications
- Ø:30 mm
- Housing material:PP
- Pore size:0.45 µm
- Cat. No.:490007-506
- Filtration area:4.8 cm²
- Membrane:PTFE
- Supplier no.:1224150
Specifications
About this item
GVS cameo syringe filters are suitable for a wide variety of applications, including HPLC sample preparation, dissolution testing, content uniformity, environmental samples, composite assays, food analysis, and Biofuel analysis.
- 33 mm housing diameter
- FLL-MLS inlet/outlet
- Non-sterile
Pure polypropylene housing, heat-sealed without the use of glue or sealants.