Pipette Racks, 4 Shelves
Supplier: Heathrow Scientific
Certificates
About this item
Protects and organizes fragile pipettes.
- Pipette sizes: 0.1 to 50 ml
ABS plastic racks have cut-out windows and include two removable attachments that hold pipette bulbs, fillers, and pumps. Magnetized version has strong magnets on both sides and mounts securely on fume hoods, file cabinets, and refrigerators. Clear version does not have attachments.
Available in acrylic or ABS plastic. Angled four shelved compartments keep your 0.1 to 50 ml pipettes or cans within easy reach while preventing damage and clutter. Crystal-clear, heavy-duty acrylic model allows viewing from any angle.