Orders LinkContactUsLinkComponent
Measuring (Mohr) Pipettes, Class A
Measuring (Mohr) Pipettes, Class A
Supplier:   Heathrow Scientific
Measuring (Mohr) Pipettes, Class A
Certificates
Measuring (Mohr) Pipettes, Class A
Measuring (Mohr) Pipettes, Class A
3 Options Available Below
Product Details & Documents
Mohr Pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of liquid handling needs.
  • Made out of high-quality borosilicate glass
  • Wide range of volume sizes to choose from
  • Reusable
  • Calibrated to deliver
They also have graduation markings from the baseline above the tip (tempered) and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges.
Product Family Options
Product Information
  • Capacity
    Band color
    Graduation Interval
    Tolerance
    Availability
    Price
  • 10 ml
    Orange
    0.10 ml
    ±0.03 ml
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $385.32
    $385.32
    Case of 72
       $385.32
    Supplier Number: 2010A-10
  • 25 ml
    White
    0.10 ml
    ±0.10 ml
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $1,082.98
    $1,082.98
    Case of 72
       $1,082.98
    Supplier Number: 2010A-25
  • 5 ml
    Blue
    0.10 ml
    ±0.02 ml
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $366.06
    $366.06
    Case of 72
       $366.06
    Supplier Number: 2010A-5