SP Bel-Art Pipette Pump Pipettors, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Provides safe, accurate and trouble-free pipetting with simple, one-handed operation.
- Pipette fits smoothly into its flexible tapered chuck
- Thumbwheel on side is rotated for precision aspirating or dispensing
- Plunger may be pressed for quick emptying
- Resistant to acids and alkalis
Disassemble all parts for cleaning and before autoclaving at 121 °C (250 °F) for 20 minutes; chuck should not be autoclaved.
Ordering information: Replacement silicone rubber chuck and washers are available must be ordered separately.