GeneMate Racked Barrier Pipette Tips, Sterile
: GeneMate
Thin wall crown design provides optimum seal and universal fit on all standard designed single and multichannel pipettors.
- More flexible than your traditional, rigid pipette tip - better ergonomic design
- Manufactured using both recycled and recyclable materials
- Ink jet identification used to preserve recyclability
- Fully recyclable trays and racks
- Graduation marks on all tips for quick volume reference
- Certified-Free of detectable DNase, RNase, and human DNA
Dense polyethylene barrier protects against aerosol and liquid contamination and provides the highest levels of purity. All trays are clearly labeled with volume and part number.