Multi-Channel Reagent Reservoir
490007-612
- Capacity:5/50 ml
- Description:Dual solutions reservoir with lids
- Cat. No.:490007-612
- Supplier no.:HS20821A
Multi-functional reagent reservoir has a single compartment 50 mL basin on one side and 12 slotted 5 ml basins on the other side.
- Angled wells guide pipette tips toward a center position to help avoid cross contamination and waste
- Individual 5 mL basin wells numerical imprinted to identify samples
- Cover included to prevent evaporation and contamination
- Made of polypropylene
Dimensions: 12.5×5×3.7 cm (4.9×2×1.5").