CappAero™ Premium Multi-channel Pipettes
Supplier: OXFORD LAB PRODUCTS INC
Certificates
About this item
Engineered for maximum performance and comfort.
- Light weight great ergonomic fit in hand
- Low plunger forces
- Fully autoclavable
Double O-ring seal on shaft for easy mounting and ejection of the tips. Stainless steep piston for excellent accuracy and precision. Virtually no maintenance required- No annual seals and O-rings to replace. Aluminum alloy shaft is virtually unbreakable- No more shaft replacements. Interchangeable control knobs for rapid volume setting changes. Calibrate your Capp Pipettes with no down time. 3-year warranty. The 30 to 300 µl, variable from 125-3 µl only. Below 125 µl please use fixed volume controllers. CappAero is a trademark of Capp A/S.