ContactUsLinkComponent
Inoculation Loops, 1 µl, with Calibration Certificate, Globe Scientific
Inoculation Loops, 1 µl, with Calibration Certificate, Globe Scientific
Catalog # 89209-784
Supplier:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Inoculation Loops, 1 µl, with Calibration Certificate, Globe Scientific
Catalog # 89209-784
Supplier:  Globe Scientific
Supplier Number:  2821
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Green
  • Size:
    1 µL
  • Cat. No.:
    89209-784
  • Supplier no.:
    2821
Specifications
About this item
These 1uL disposable inoculation loops offer a convenient and safe way of streaking culture media
These loops are enclosed in peel-apart packages, with each pack containing 20 loops. All loops are sterilized by gamma radiation.
Molded from a special grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Certifications: Each case comes with a Certificate of Calibration.