This antibody reacts with a 180 kDa protein, known as Flt-1 or vascular endothelial growth factor receptor 1 (VEGFR1). Flt-1 is a cell membrane receptor kinase. Flk-1 has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.
Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 95025-024
- 99991-328
- Antigen Symbol:
- FLT1
- FLT1
- Antigen Name:
- FMS-Related Tyrosine Kinase 1
- FMS-Related Tyrosine Kinase 1
- Antigen Synonyms:
- FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
- FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
- Clonality:
- Polyclonal
- Polyclonal
- Concentration:
- Concentrated
- Concentrated