Anti-Thyroid Peroxidase Mouse Monoclonal Antibody [clone: MoAb47]
Supplier: Diagnostic Biosystems
Certificates
About this item
Thyroid peroxidase (TPO) is a transmembrane protein of 107kDa containing a heme prosthetic group. It is present as a dimer on the apical surface of thyroid follicular cells. TPO is the primary enzyme involved in thyroid hormone synthesis. Malignant thyroid tumors exhibit an anomaly in TPO resulting in lower affinity for anti-TPO. This antibody may aid in the differentiation between benign and malignant thyroid tumors.
Type:
Antigen: Thyroid Peroxidase
Clonality: Monoclonal
Clone: MoAb47
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- 95024-844
- 99990-772
- Antigen Symbol:
- TPO
- TPO
- Antigen Name:
- Thyroid Peroxidase
- Thyroid Peroxidase
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- MoAb47
- MoAb47
- Host:
- Mouse
- Mouse
- Concentration:
- Concentrated
- Concentrated