Anti-Thymidine Phosphorylase Mouse Monoclonal Antibody [clone: P-GF.44C]
This antibody is specific to a 55 kDa protein known as platelet-derived endothelial growth factor (PD-ECGF), or thymidine phosphorylase (TP). High levels of TP/PD-ECGF are observed in cancer patients. High intracellular levels of TP/PD-ECGF are associated with increased sensitivity to fluoropyrimidine chemotherapeutic agents.
Type:
Antigen: Thymidine Phosphorylase
Clonality: Monoclonal
Clone: P-GF.44C
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
- Catalog No:
- 99990-552
- 95024-834
- Antigen Symbol:
- Thymidine Phosphorylase
- Thymidine Phosphorylase
- Antigen Name:
- Thymidine Phosphorylase
- Thymidine Phosphorylase
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- P-GF.44C
- P-GF.44C
- Host:
- Mouse
- Mouse
- Concentration:
- Concentrated
- Concentrated