This antibody recognizes human retinoblastoma gene protein. Rb gene, a prototype of tumor suppressor genes, has been associated with development and/or progression of bladder cancer as well as sarcoma and small cell lung cancer. Functional Rb gene loss has been reported to occur in bladder cancer and osteosarcoma. A significantly poorer tumor-free survival rate was noted for those patients who had a tumor with an altered Rb protein with or without vascular invasion.
Type:
Antigen: Retinoblastoma
Clonality: Monoclonal
Clone: 1F8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 99990-998
- 99990-440
- 95024-810
- Antigen Symbol:
- Retinoblastoma
- Retinoblastoma
- Retinoblastoma
- Antigen Name:
- Retinoblastoma
- Retinoblastoma
- Retinoblastoma
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 1F8
- 1F8
- 1F8
- Host:
- Mouse
- Mouse
- Mouse
- Concentration:
- Ready to use
- Concentrated
- Concentrated