Orders LinkContactUsLinkComponent
Liquid Sampling Kit, Dynalon
Liquid Sampling Kit, Dynalon
Liquid Sampling Kit, White
Catalog # 10053-758
Supplier:   Dynalon
Supplier #:  107035-3000
Liquid Sampling Kit, Dynalon
Catalog # 10053-758
Supplier:  Dynalon
Liquid Sampling Kit, Dynalon
Liquid Sampling Kit, Dynalon
Liquid Sampling Kit, White
Price
$509.32
Log in for availability
Each
Product Details & Documents
These versatile modular design plastic liquid sampling kits will meet all of your needs
Kit components are manufactured of high density polyethylene (HDPE) and polypropylene (PP). Their threaded connections allow quick and easy use. The Liquid Sampling Kit includes a sampler component with 100ml and 500ml polypropylene bottle plus a 1.5', 3' and 6' HDPE handle.
Whether sampling from drums, tanks, or rail cars, taking water samples or dipping chemicals from a bath we have a kit configuration to meet your liquid sampling application requirements.
Weight: 5.1 lbs.
Certifications: FDA Compliant.
Ordering information: Liquid Sampling Kit components are interchangeable. Component are also sold as separate items (See 10053-764, -766, -768, -750 and -752). Kits and their components are available as one each.
Specifications
  • Description:
    Liquid Sampling Kit, White
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Handle, HDPE, 1.5 Ft., White, 18L x 2W", 0.4 lb.
    Handle, HDPE, 1.5 Ft., White, 18L x 2W", 0.4 lb.
    10053-764
    Log in for availability
    Each - $70.21
Product Information
  • Catalog #
    Availability
    Price
  • Handle, HDPE, 3 Ft., White, 36L x 5W", 0.7 lb.
    Handle, HDPE, 3 Ft., White, 36L x 5W", 0.7 lb.
    10053-766
    Log in for availability
    Each - $88.35
Product Information
  • Catalog #
    Availability
    Price
  • Handle, HDPE 6 Ft., White, 72L x 2W", 2.5 lbs.
    Handle, HDPE 6 Ft., White, 72L x 2W", 2.5 lbs.
    10053-768
    Log in for availability
    Each - $114.27
Product Information
  • Catalog #
    Availability
    Price
  • Sampler (HDPE) with Bottle (PP), 100 ml, Natural, 7L x 2W", 0.5 lb.
    Sampler (HDPE) with Bottle (PP), 100 ml, Natural, 7L x 2W", 0.5 lb.
    10053-750
    Log in for availability
    Each - $138.07
Product Information
  • Catalog #
    Availability
    Price
  • Sampler (HDPE) with Bottle (PP), 500 ml, Natural, 10L x 3W", 1 lb.
    Sampler (HDPE) with Bottle (PP), 500 ml, Natural, 10L x 3W", 1 lb.
    10053-752
    Log in for availability
    Each - $201.62