Specifications
- Description:Reticulum Stain
- Cat. No.:95027-218
- No. of reactions:100
- Supplier no.:KT 031
Specifications
About this item
This kit stains reticulum fibers in paraffin tissue sections
The reticulum fibers stain gray to black, while the nuclei and background stain pink to rose.