ContactUsLinkComponent
Fontana Masson Kit, Diagnostic Biosystems
Fontana Masson Kit, Diagnostic Biosystems
  95027-188
 :  
Fontana Masson Kit, Diagnostic Biosystems
  95027-188
 :  KT014
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Fontana Masson Kit
  • Cat. No.:
    95027-188
  • No. of reactions:
    100
  • Supplier no.:
    KT 014
This kit stains argentaffin cells and melanin in paraffin tissue sections
The argentaffin cells and melanin granules stain brown to black, while the nuclei stain pink.