Differential Quick Stain, Electron Microscopy Sciences
About this item
This kit is a Modified Giemsa Stain kit for quick turnaround results. It may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms.
- The stain offers results in 15 seconds
- Comes with a fixative for air dried cell suspensions and or touch preparation slides
Used for fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies.
: Certificates of analysis are available.
: The Kit comes complete with 3 solutions and is available in 3 sizes.
2 Options Available Below
Product Details & Documents
This kit is a Modified Giemsa Stain kit for quick turnaround results. It may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms.
- The stain offers results in 15 seconds
- Comes with a fixative for air dried cell suspensions and or touch preparation slides
Used for fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies.
: Certificates of analysis are available.
: The Kit comes complete with 3 solutions and is available in 3 sizes.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 102773-654Differential Quick Stain KitSupplier Number: 26096-25Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 102773-656Differential Quick Stain KitSupplier Number: 26096-50
- Discontinued-Supplier Number: -