ContactUsLinkComponent
The Grid Dot Array, Electron Microscopy Sciences
The Grid Dot Array, Electron Microscopy Sciences
Catalog # 102773-622
The Grid Dot Array, Electron Microscopy Sciences
Catalog # 102773-622
Supplier Number:  68073-R76
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Grid Dot Array
  • Cat. No.:
    102773-622
  • Supplier no.:
    68073-R76
Specifications
About this item
The ideal product for testing image area, distortion, field flatness and other parameters in optical and imaging systems.
Green Float glass, high reflective Chrome image. The three array areas give options for different magnifications or field size. The R76, a unique product with three dot arrays to suit different magnification or image areas.
Features 3 image areas, with dot size and pitch different in each area:
12 x 9 array of 1 mm dots at 5 mm pitch
16 x 12 array of 0.5 mm dots at 2 mm pitch
24 x 18 array of 0.2 mm dots at 1 mm pitch