Omniprobe 4 Post, Electron Microscopy Sciences
Product Details & Documents
The 4 post has copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations, two with vertical bar attachment surfaces and two with 'V'shaped alighment surfaces. Sides have lower profile for easier access to outermost posts.
Packaging: Packaged in plastic vials.
Specifications
- Ø:3 mm
- Description:4 Post
- Thickness:30 µm