Specifications
- Description:3 Post Copper Liftout Grids
- Cat. No.:102773-586
- Supplier no.:75964-08
Specifications
About this item
The 3 post copper lift-out grids are specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both vertical bar and 'V' shaped attachment surfaces. These grids have a shallower downset and slightly wider center post.