Specifications
- Description:PFPE 501 Grease
- Size:100 g
- Cat. No.:102773-572
- Supplier no.:60708
Specifications
About this item
Why Use Apiezon PFPE 501 Grease?
PFPE 501 is a chemically inert, high temperature and extreme pressure lubricant. With an upper operating temperature of 250°C and vapour pressure of 1.3 x 10-12 at 25°C, it is robust, versatile and can be used for sealing and lubricating under the most extreme operating conditions. This includes environments where aggressive chemicals and strong oxidising agents are regularly used.
Why Use Apiezon PFPE 501 Grease?
PFPE based
High temperature lubricant
Used in oxygen rich environments
Ultra high vacuum
Chemically inert
Extreme Pressure
Wide temperature range
Benefits:
Extended equipment life
High temperature lubricant
Reduced equipment downtime
Lower service costs
Life-time lubrication
Guaranteed Apiezon quality
Expert technical support
Benefits:
Extended equipment life
High temperature lubricant
Reduced equipment downtime
Lower service costs
Life-time lubrication
Guaranteed Apiezon quality
Expert technical support