Orders LinkContactUsLinkComponent
Peak ABC Loupe, Electron Microscopy Sciences
Peak ABC Loupe, Electron Microscopy Sciences
ABC Loupe
Catalog # 102773-570
Supplier #:  68160
Peak ABC Loupe, Electron Microscopy Sciences
Catalog # 102773-570
Peak ABC Loupe, Electron Microscopy Sciences
Peak ABC Loupe, Electron Microscopy Sciences
ABC Loupe
Price
$869.45
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Peak ABC Loupe is a precision folding loupe, consisting of A lens (12X), B lens (3X) and C lens (6X).
With these three lenses combined, seven kinds of magnifying power are available. The lens is made from high quality glass optics and the body is made of aluminum. The magnification desired will be available from the combination table shown on the back of the loupe.
Specifications
  • Magnification:
    3×, 6×, 9×, 12×, 14×, 16×, 18×
  • Description:
    ABC Loupe