Micro Sample Tubes, Electron Microscopy Sciences
Product Details & Documents
Suitable for use in the mini centrifuge.
Polypropylene tubes with a captive plug, they are steam autoclavable at 121 °C (250 °F).
Specifications
- Description:Micro Sample Tubes, 1.5ul